Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
IP, WB
Citations:
4
biological source
rabbit
Quality Segment
conjugate
unconjugated
antibody form
affinity purified immunoglobulin
antibody product type
primary antibodies
clone
polyclonal
species reactivity
human
technique(s)
immunoprecipitation (IP): 2- 5 μg/mg, western blot: 1:2,000- 1:10,000
accession no.
NP_005643.1
shipped in
wet ice
storage temp.
2-8°C
target post-translational modification
unmodified
Gene Information
rabbit ... TRF2(7014)
General description
The telomeric repeat binding factor 2 (TRF2 ) gene codes for a component of shelterin. The encoded protein is dimer containing four domains : B domain at the N-terminal, the TRF homology domain, the hinge domain and the MYB/SANT (proto-oncogene, transcription factor telobox) domain at C-terminal.
Immunogen
The epitope recognized by PLA0144 maps to a region between residue 375 and 425 of human Telomeric Repeat Binding Factor 2 using the numbering given in entry NP_005643.1 (GeneID 7014).
Biochem/physiol Actions
TRF2 (telomeric repeat binding factor 2) is responsible for telomere stability by the insertion of the 3′ overhang. TRF2 offers protection to DNA from being wrongly recognized by the DNA damage repair systems. Loss of TRF2 function induces DNA repair system resulting in apoptosis by causing cell cycle arrest. Upregulation of the TRF2 gene is observed in cancer and thus concluded to contribute to tumor progression.
Physical form
Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide
Other Notes
Telomeric repeat binding factor (TRF2) is a ubiquitously expressed protein that localizes at chromosome ends during all phases of the cell cycle and binds directly to TTAGGG repeats. TRF2 functions to protect telomeres as being perceived as sites of DNA damage. TRF2 interacts with several proteins at telomeres, such as Rap1, Mre11, Rad50, and NBS1.
Disclaimer
Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.
Still not finding the right product?
Try our Product Selector Tool to narrow your options
Storage Class
12 - Non Combustible Liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.