Skip to Content
MilliporeSigma

PLA0118

Rabbit anti-Phospho RPS6 (S235/236) Antibody, Affinity Purified

Powered by Bethyl Laboratories, Inc.

Synonym(s):

40S ribosomal protein S6, S6, phosphoprotein NP33

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.41
UNSPSC Code:
12352203
Conjugate:
unconjugated
Clone:
polyclonal
Application:
WB
Citations:
8
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

rabbit

Quality Segment

conjugate

unconjugated

antibody form

affinity purified immunoglobulin

antibody product type

primary antibodies

clone

polyclonal

species reactivity

human

technique(s)

western blot: 1:2,500-1:10,000

accession no.

NP_001001.2

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

phosphorylation (pSer235/236)

Gene Information

human ... RPS6(6194)

Immunogen

Immunogen for PLA0118 was a phosphorylated synthetic peptide, which represented a portion of human Ribosomal Protein S6 (GeneID 6194) around serine 235 and serine 236 according to the numbering given in entry NP_001001.2.

Physical form

Tris-buffered Saline containing 0.1% BSA containing 0.09% Sodium Azide

Other Notes

Ribosomal protein S6 (RPS6) is a component of the 40S ribosomal subunit. Ribosomes are composed of a small 40S subunit and a large 60S subunit that function to catalyze protein synthesis. RPS6 is phosphorylated by many cellular kinases including ribosomal protein S6 kinase that is regulated via the mammalian target of rapamycin (mTOR) pathway. RPS6 has been demonstrated to be important in regulating cell size, glucose homeostasis, and translational control of mRNA.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.


Still not finding the right product?

Try our Product Selector Tool to narrow your options


Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library