Skip to Content
MilliporeSigma

PIR00

Inhibitor Resistant Genotyping PCR ReadyMix

for Bio-Rad, Cepheid, Eppendorf, Illumina, Corbett, and Roche systems

Synonym(s):

Inhibitor Resistant Genotyping PCR ReadyMix

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.55
UNSPSC Code:
41106300
Technique(s):
RT-qPCR: suitable, qPCR: suitable
Usage:
sufficient for 1250 reactions, sufficient for 250 reactions, sufficient for 5000 reactions
Storage temp.:
−20°C
Detection method:
probe-based
Shipped in:
dry ice
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

liquid

usage

sufficient for 1250 reactions, sufficient for 250 reactions, sufficient for 5000 reactions

feature

Difficult Templates/Specialty Enzymes PCR, dNTPs included, hotstart

storage condition

protect from light

technique(s)

RT-qPCR: suitable, qPCR: suitable

color

light blue

input

crude DNA

compatibility

Bio-Rad CFX384, Bio-Rad CFX96, Bio-Rad MJ Chromo4, Bio-Rad MJ Opticon 2 , Bio-Rad MJ Opticon, Bio-Rad MiniOpticon, Cepheid SmartCycler, Eppendorf® Mastercycler ep realplex , Eppendorf® Mastercycler ep realplex2 S, Illumina Eco qPCR, Qiagen Corbett Rotor-Gene 3000, Qiagen Corbett Rotor-Gene 6000, Qiagen Corbett Rotor-Gene Q, Roche LightCycler 480

detection method

probe-based

shipped in

dry ice

storage temp.

−20°C

General description

Inhibitor Resistant Genotyping PCR Readymix is a 2X concentrated ready-to-use master mix for PCR amplification of DNA templates that overcomes inhibitors often present in crude samples extracted from environmental specimens, plant tissues, or animal tissues. It is a versatile and robust PCR reagent that provides highly specific and efficient amplification for genotyping sequence variants including allelic discrimination of single nucleotide polymorphisms (SNPs) using a variety of fluorogenic probe chemistries, including TaqMan® hydrolysis probes. Inhibitor Resistant Genotyping PCR Readymix contains all required reaction components, except primers, probes, and DNA template. The light blue color of the inert tracer dye simplifies reaction assembly in white, or clear, plates and helps to minimize pipetting or mixing errors. It does not interfere with reporter dye fluorescence or affect the stability of the product.

A key component of Inhibitor Resistant Genotyping PCR ReadyMix is an ultra-pure, highly processive thermostable DNA polymerase that is combined with high avidity monoclonal antibodies. This proprietary polymerase mix is highly resistant to PCR inhibitors and provides an extremely stringent automatic hot-start allowing reaction assembly, and temporary storage, at room temperature prior to PCR amplification.

Application

Different real-time PCR systems employ different strategies for normalization of fluorescent signals and correction of well-to-well optical variations. It is critical to match the appropriate qPCR reagent to your specific instrument.KiCqStart® One-Step Probe RT-qPCR ReadyMix does not contain an internal reference dye.
Inhibitor Resistant Genotyping PCR ReadyMix has been used as a component of the reaction mixture for detection of leucoanthocyanidin reductase(PaLAR3) alleles in a genotyping assay and multiplex PCR for amplification of microsatellite.

Features and Benefits

  • Assay results in as little as 33 minutes
  • Highly efficient and sensitive real-time PCR results
  • Little/no optimization required
  • Compatible with both fast and conventional PCR cycling protocols

Other Notes

2X reaction buffer containing optimized concentrations of MgCl2, dNTPs (dATP, dCTP, dGTP, dTTP), hot-start DNA polymerase, insert blue qPCR dye, and stabilizers.

packaging:
250 reactions* = 2 X 1.25 mL tubes
1250 reactions* = 10 X 1.25 mL tubes
5000 reactions* = 1 X 50 mL tube
*number of reactions based on a 20uL volume
Compatible instruments include:
  • Bio-Rad® CFX384
  • Bio-Rad CFX96
  • Bio-Rad MiniOpticon
  • Bio-Rad/MJ Chromo4
  • Bio-Rad/MJ Opticon 2
  • Bio-Rad/MJ Opticon
  • Cepheid SmartCycler®
  • Eppendorf Mastercycler® ep realplex
  • Eppendorf Mastercycler ep realplex2 s
  • Illumina Eco qPCR
  • Qiagen/Corbett Rotor-Gene® 3000
  • Qiagen/Corbett Rotor-Gene 6000
  • Qiagen/Corbett Rotor-Gene Q
  • Roche LightCycler® 480

Legal Information

Bio-Rad is a registered trademark of Bio-Rad Laboratories, Inc.
CFX384 is a trademark of Bio-Rad Laboratories, Inc.
CFX96 is a trademark of Bio-Rad Laboratories, Inc.
Chromo4 is a trademark of Bio-Rad Laboratories, Inc.
Eppendorf is a registered trademark of Eppendorf SE
KiCqStart is a registered trademark of QIAGEN Beverly Inc.
LightCycler is a registered trademark of Roche
Mastercycler is a registered trademark of Eppendorf SE
MiniOpticon is a trademark of Bio-Rad Laboratories, Inc.
Opticon is a trademark of Bio-Rad Laboratories, Inc.
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
Rotor-Gene is a registered trademark of Qiagen GmbH
SmartCycler is a registered trademark of Cepheid, Inc.
TaqMan is a registered trademark of Roche Molecular Systems, Inc.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library