Skip to Content
MilliporeSigma

P9769

Decaethylene glycol mono­dodecyl ether

nonionic surfactant

Synonym(s):

C12E10, Polyoxyethylene (10) lauryl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C32H66O11
CAS Number:
Molecular Weight:
626.86
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

form

semisolid (with melting point ~24 °C.)

mol wt

626.86 g/mol

SMILES string

CCCCCCCCCCCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCOCCO

InChI

1S/C58H118O24/c1-2-3-4-5-6-7-8-9-10-11-13-60-15-17-62-19-21-64-23-25-66-27-29-68-31-33-70-35-37-72-39-41-74-43-45-76-47-49-78-51-53-80-55-57-82-58-56-81-54-52-79-50-48-77-46-44-75-42-40-73-38-36-71-34-32-69-30-28-67-26-24-65-22-20-63-18-16-61-14-12-59/h59H,2-58H2,1H3

InChI key

IEQAICDLOKRSRL-UHFFFAOYSA-N

Application

Decaethylene glycol mono­dodecyl ether has been used as a non-ionic surfactant to study its characterization and interactions with bovine serum albumin (BSA) and to assess their complexes.
Decaethylene glycol mono­dodecyl ether has been used in a study to assess diffusion of proteins and nonionic micelles in agarose gels. It has also been used in a study to investigate properties of anti-inflammatory resolvins derived from docosapentaenoic acid.


signalword

Warning

Storage Class

11 - Combustible Solids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves

pictograms

Exclamation mark

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library