Skip to Content
MilliporeSigma

P8241

Nunc® 96 DeepWell plate, non-treated

size 96 wells, U-bottom natural polypropylene wells, maximum volume 1.3 mL, non-sterile

Synonym(s):

96 deepwell culture plate, 96 deepwell plate, Nunc 260252, cell cultivation plate, culture plate, multiwell plate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.22
UNSPSC Code:
41122107
Material:
U-bottom natural polypropylene wells, natural polypropylene , polypropylene
Size:
96 wells
Sterility:
non-sterile
Binding type:
non-treated surface
Feature:
lid: no
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

U-bottom natural polypropylene wells, natural polypropylene , polypropylene

sterility

non-sterile

feature

lid: no

packaging

case of 50 ea (internal packs of 5)

manufacturer/tradename

Nunc 260252

maximum volume

1.3 mL

plate L × W

128 mm × 86 mm

size

96 wells

well maximum volume

1.3 μm

working volume

1 mL

color

natural

suitability

suitable for (sample collection, storage, combinatorial chemistry and library applications)

binding type

non-treated surface

General description

Optimize storage capacity and improve mixing using Nunc® 1.3 and 2.0mL DeepWell™ plates with shared-wall technology. Ideal for sample collection, storage, combinatorial chemistry and library applications, and resistant to most chemicals, solvents and alcohols used in combinatorial chemistry.




The large well diameters offer ideal access and excellent mixing characteristics, and the U shaped well enhances compatibility with the magnetic bead extraction. The V shaped well bottom ensures low dead-volume and high sample recovery during extraction.




Manufactured in a cleanroom facility or clean environment to ensure molecular grade quality, polypropylene plates provides excellent solutions for sample storage and assay set-up, allowing dilutions and aliquots to be handled, stored, or transported easily. The Virgin medical grade resin passes requirements for USP <661.1> and USP <665> reports have been completed for both CapMat sealed and Heat Sealed plates.

Application

  • Combinatorial chemistry and library applications

Features and Benefits

  • Unique low design with shared well technology for increased well volume1.3 and 2.0 mL volume capacity
  • Optimum storage capacity and improved mixing
  • Round well shape reduces liquid retention
  • Well cap available for safe storage – minimal protrusion into each well
  • Ideal for sample collection, storage, combinatorial chemistry and library applications
  • Resistant to most chemicals, solvents and alcohols used in combinatorial chemistry
  • Widely used for bacterial and yeast growth
  • Easy-to-use with automatic sample handling instruments
  • Sealable with well caps and sealing tape

Legal Information

DeepWell is a trademark of Thermo Fisher Scientific or its subsidiaries
Nunc is a registered trademark of Thermo Fisher Scientific or its subsidiaries


Still not finding the right product?




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library