Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

P8004

Sigma-Aldrich

5α-Pregnan-3β-ol-20-one 3β-acetate

≥98%

Synonym(s):

3β-Hydroxy-5α-pregnan-20-one acetate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H36O3
CAS Number:
Molecular Weight:
360.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

SMILES string

C[C@@]12[C@](CC[C@@H]2C(C)=O)([H])[C@]3([H])CC[C@@]4([H])C[C@@H](OC(C)=O)CC[C@]4(C)[C@@]3([H])CC1

InChI

1S/C23H36O3/c1-14(24)19-7-8-20-18-6-5-16-13-17(26-15(2)25)9-11-22(16,3)21(18)10-12-23(19,20)4/h16-21H,5-13H2,1-4H3/t16-,17?,18?,19?,20?,21?,22?,23?/m0/s1

InChI key

GFHOQCXDABGYAL-ZVKOYPPWSA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Anna A Rybczynska et al.
European journal of nuclear medicine and molecular imaging, 36(7), 1167-1175 (2009-02-28)
Sigma receptors are implicated in memory and cognitive functions, drug addiction, depression and schizophrenia. In addition, sigma receptors are strongly overexpressed in many tumours. Although the natural ligands are still unknown, steroid hormones are potential candidates. Here, we examined changes
Ruoyu Wen et al.
PloS one, 7(9), e44681-e44681 (2012-09-13)
In most eukaryotes that have been studied, the telomeres cluster into a bouquet early in meiosis, and in wheat and its relatives and in Arabidopsis the centromeres pair at the same time. In Arabidopsis, the telomeres do not cluster as

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service