Skip to Content
MilliporeSigma

P7543

PARAFILM® M

roll size 2 in. × 250 ft

Synonym(s):

Covering Film, Sealing Film

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NB.35
UNSPSC Code:
41122410
Manufacturer/tradename:
Parafilm PM992
Material:
film (thermoplastic)
W × l:
5 cm × 76 m
Sterility:
non-sterile
Packaging:
pack of 1 ea
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


material

film (thermoplastic)

sterility

non-sterile

packaging

pack of 1 ea

manufacturer/tradename

Parafilm PM992

parameter

-45-+50 °C temp. range

W × L

5 cm × 76 m

roll size

2 in. × 250 ft

General description

PARAFILM stretches up to 200% and clings even around irregular shapes and surfaces. The sealing film is free of plasticizers and consists primarily of polyolefins and paraffin waxes. PARAFILM M is resistant up to 48 hours against many polar substances, eg saline solutions, inorganic acids and alkaline solutions.

Protect the integrity of research applications with a tight moisture-barrier that holds a strong seal and doesn′t release from glassware, despite off-gassing
Reduce the risk of tampered data with a covering that is odorless, colorless and semi-transparent
Prevents moisture and volume loss due to material composition and tight seal
Flexible and self-sealing material allows it to cling around irregular shapes and surfaces
Stretches more than 200% of original length
Resistant up to 48 hours against many polar substances such as saline solutions, inorganic acids and alkaline solutions

Permeability

Oxygen: (ASTM 1927-98) 150 cc/m² d at 23°C and 50% RH
Carbon Dioxide: (Modulated IR Method) 1200 cc/m² d at 23°C and 0% RH
Water Vapor: (ASTM F1249-01)
Flat: 1 g/m² d at 38°C and 90% RH
Creased: 1 g/m² d at 38°C and 90% RH

Effects of Common Reagents
Potassium Permanganate
5%: No apparent effect except permanent dark brown coloration in 18 hours
0.1%: Same as 5% except slightly less color
Iodine solution (0.1N)
No effect except staining brown in 18 hours
Ethyl Alcohol (95%)
No apparent effect except some face whitening in 24 hours
No apparent effect in 24 hours:
Hydrochloric Acid conc. (12N) dil. (5N)
Sulphuric Acid conc. (36N) dil. (5N)
Nitric Acid conc. (16N) dil. (5N)
Sodium Hydroxide conc. (22%)
Ammonium Hydroxide conc. (28% NH<sub>3</sub>)
Salt (NaCl) solution (20%)
Isopropyl Alcohol (99%)

Not recommended for use with chlorinated, non-polar aliphatic and aromatic solvents
Film becomes soft and sticky at about 130° to 150°F (54° to 66°C)

Legal Information

Parafilm is a registered trademark of Bemis Company, Inc.


Still not finding the right product?

Explore all of our products under PARAFILM® M




Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library