Skip to Content
MilliporeSigma

P7443

L-α-Phosphatidylcholine

from soybean, ≥99% (TLC), lyophilized powder

Synonym(s):

1,2-Diacyl-sn-glycero-3-phosphocholine, 3-sn-Phosphatidylcholine, L-α-Lecithin, Azolectin, PC

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352211
NACRES:
NA.25
EC Number:
232-307-2
Beilstein/REAXYS Number:
5209585
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

soybean

Quality Segment

assay

≥99% (TLC)

form

lyophilized powder

functional group

phospholipid

lipid type

phosphoglycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

[P](=O)([O-])(OC[C@H](OC(=O)CCCCCCC\C=C/C\C=C/CCCCC)COC(=O)CCCCCCCCCCCCCCC)OCC[N+](C)(C)C

InChI

1S/C42H80NO8P/c1-6-8-10-12-14-16-18-20-21-23-25-27-29-31-33-35-42(45)51-40(39-50-52(46,47)49-37-36-43(3,4)5)38-48-41(44)34-32-30-28-26-24-22-19-17-15-13-11-9-7-2/h14,16,20-21,40H,6-13,15,17-19,22-39H2,1-5H3/b16-14-,21-20-/t40-/m1/s1

InChI key

JLPULHDHAOZNQI-ZTIMHPMXSA-N

General description

L-α-Phosphatidylcholine (PC) is an asymmetric molecule with glycerol backbone and occurs naturally in lecithin of egg yolk. It is a phospholipid and major choline source in mammalian cells. It is crucial for very low-density lipoprotein secretion in hepatocytes. It is essential for the acetylcholine synthesis functions to nourish the brain. PC plays a crucial role in lipid and lipoprotein metabolism. Deficiencies of enzymes in PC biosynthetic pathway leads to an array of metabolic disorder. Intracellular levels of hepatic PC to phosphatidylethanolamine (PE) ratio contributes to pathophysiology non-alcoholic fatty liver disease (NAFLD). It has anti-inflammatory functionality and is effective in treating inflammatory bowel disease. PC administration therapy is effective for treating benign symmetric lipomatosis.

Application

L-α-Phosphatidylcholine has been used:
  • for liposome preparation
  • as a component of stabilizer solution for preparing dispersed curcuminoid submicron particle (CSP)
  • as a component of serum free substitute for whole blood cells

Biochem/physiol Actions

A major structural phospholipid in brain, comprising approx. 15% of total lipid; primarily localized to gray matter.

Packaging

Packaged under Argon

Preparation Note

Prepared by a modification of the procedure of Singleton, W.S., et al., J. Am. Oil Chem. Soc., 42, 53 (1965).


Still not finding the right product?

Explore all of our products under L-α-Phosphatidylcholine


Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library