Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C11H8O3
CAS Number:
Molecular Weight:
188.18
NACRES:
NA.79
PubChem Substance ID:
UNSPSC Code:
12352205
EC Number:
207-569-6
MDL number:
biological source
Plumbago indica
Quality Segment
form
powder
color
faint orange to dark orange
mp
76-78 °C (lit.)
storage temp.
−20°C
SMILES string
CC1=CC(C2=C(C1=O)C=CC=C2O)=O
InChI
1S/C11H8O3/c1-6-5-9(13)10-7(11(6)14)3-2-4-8(10)12/h2-5,12H,1H3
InChI key
VCMMXZQDRFWYSE-UHFFFAOYSA-N
General description
Plumbagin is a bioactive naphthoquinone present majorly in Plumbago indica L. It is a quinoid and is also derived from the roots of Plumbago zeylanica roots.
Application
Plumbagin from Plumbago indica has been used:
- as a reactive oxygen species agent (ROS) to induce cytotoxicity in mouse embryonic fibroblasts
- as an oxidative stress inducer to generate superoxide anion in Saccharomyces cerevisiae
- as a reference standard in thin layer chromatography and in spectrophotometric analysis for quantification of plumbagin in Plumbago auriculate samples
Biochem/physiol Actions
Exhibits cytotoxicity in rodent models of carcinogenesis and carcinoma; has antifungal, antiviral, and antibacterial action.
Plumbagin exhibits various pharmacological activities including antimicrobial, anticancer, anti-atherosclerotic, antidiabetic, anti-inflammatory, hypolipidemic, and neuroprotective activities. It inhibits the signal transducer and activator of transcription 3 (STAT3) signaling and halts the proliferation of esophageal squamous cell carcinoma (ESCC). Plumbagin elicits protective antioxidative functionality in 4-nitroquinoline-N-oxide (NQNO) induced stress in lymphoma. Plumbagin in a nanoemulsion formulation has antiproliferative effect towards prostate cancer.
signalword
Danger
hcodes
Hazard Classifications
Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
target_organs
Respiratory system
Storage Class
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P7262-250MG | 04061833275900 |
| P7262-1G | 04061834394525 |
| P7262-100MG | 04061834394518 |
