Select a Size
| Size/SKU | Availability | Price |
|---|
About This Item
material
conical bottom natural wells, natural polypropylene , polypropylene
sterility
sterile
feature
lid: no, high flange design
packaging
case of 120 ea (internal packs of 10)
manufacturer/tradename
Nunc 249946
plate L × W
128 mm × 86 mm
size
96 wells
well maximum volume
400 μm
working volume
400 μL
binding type
non-treated surface
General description
Application
Features and Benefits
- Shared wall design increases the well volume.
- Wider wells and rounded bottom improve mixing.
- Conical bottom well for optimal sample recovery.
- Black and white plates available for fluorescent- and luminescent-based applications.
- Low binding polypropylene for homogeneous assays and storage.
- Compatible with robotics and automated systems.
- Polypropylene resin is resistant to most chemicals, solvents and alcohols.
- Minimal binding of molecules with hydrophilic moities.
- Non-toxic.
- Products come without lids.
Legal Information
1 of 1
This Item | |||
|---|---|---|---|
| manufacturer/tradename Nunc 249946 | manufacturer/tradename Nunc 249944 | manufacturer/tradename Nunc 267334 | manufacturer/tradename Nunc 269620 |
| binding type non-treated surface | binding type non-treated surface | binding type non-treated surface | binding type non-treated surface |
| sterility sterile | sterility non-sterile | sterility sterile | sterility non-sterile |
| material conical bottom natural wells, polypropylene , natural polypropylene | material conical bottom natural wells, natural polypropylene | material conical bottom natural polypropylene wells, natural polypropylene plate | material clear polystyrene , flat bottom clear wells |
| feature high flange design, lid: no | feature lid: no, high flange design | feature lid: no, high flange design | feature lid: no, pinchbar; high flange design |
| well maximum volume 400 μm | well maximum volume 400 μm | well maximum volume 450 μm | well maximum volume 400 μm |
Still not finding the right product?
Explore all of our products under Nunc® MicroWell™ 96 well polypropylene plates
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.



