Skip to Content
MilliporeSigma

Skip To

P6744

Picric acid solution

1.3% in H2O (saturated)

Picric acid solution

Synonym(s):

2,4,6-Trinitrophenol

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(O2N)3C6H2OH
CAS Number:
Molecular Weight:
229.10
UNSPSC Code:
12171500
NACRES:
NA.47
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
423400
Concentration:
1.3% in H2O (saturated)
Form:
liquid
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

liquid

Quality Segment

concentration

1.3% in H2O (saturated)

color

yellow to very dark yellow

density

1.080 g/cm3

application(s)

hematology
histology

storage temp.

room temp

SMILES string

Oc1c(cc(cc1[N+]([O-])=O)[N+]([O-])=O)[N+]([O-])=O

InChI

1S/C6H3N3O7/c10-6-4(8(13)14)1-3(7(11)12)2-5(6)9(15)16/h1-2,10H

InChI key

OXNIZHLAWKMVMX-UHFFFAOYSA-N

General description

Picric acid is a polynitrated aromatic acid. It dissolves in water to form a solution with low pH (pH 2.0). It interacts with basic groups of proteins, forms salts and coagulates them. Fixation with picric acid gives a bright stain, but there might be a loss of nucleic acids due to the low pH of the solution.
Picric acid is anionic and has a low molecular weight.

Application

Picric acid is commonly used as a fixative and staining reagent in forensic and histological applications. It is a general cytoplasmic stain, connective tissue fiber stain, and a fixative for tissues.
  • It is a component of picrosirius red which is used to stain collagen.
  • Bouin’s fluid which also comprises picric acid is a fixative for nuclei and connective tissue.
  • It can be used as a colorimetric reagent in creatinine determination assay.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
4620924-31201.01799
concentration

1.3% in H2O (saturated)

concentration

100 μg/mL in acetonitrile

concentration

-

concentration

69%

Quality Level

200

Quality Level

100

Quality Level

-

Quality Level

300

form

liquid

form

-

form

liquid

form

liquid

storage temp.

room temp

storage temp.

2-8°C

storage temp.

-

storage temp.

2-30°C

density

1.080 g/cm3

density

-

density

1.005 g/cm3

density

1.41 g/cm3 at 20 °C

application(s)

hematology
histology

application(s)

agriculture
cleaning products
cosmetics
food and beverages
personal care

application(s)

-

application(s)

-


Still not finding the right product?

Explore all of our products under Picric acid solution


supp_hazards

Storage Class

12 - Non Combustible Liquids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

Active Filters

  1. 1 Ratings-Only Review