Skip to Content
MilliporeSigma

P5975

Amino-terminal Met-FLAG-BAP Fusion Protein

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
MDL number:
Mol wt:
~49 kDa
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


mol wt

~49 kDa

Quality Segment

shipped in

dry ice

storage temp.

−20°C

Application

Amino-terminal Met-FLAG-BAP Fusion Protein is useful for assurance of the functional integrity of ANTI-FLAGÒ M2 and M5 monoclonal antibodies in immunological procedures such as Western blotting, ELISA, immunoprecipitation, fluorescence microscopy, light microscopy and FACS.
Learn more product details in our FLAG® application portal.

Biochem/physiol Actions

Amino-terminal Met-FLAG-BAP control protein is a 468 amino acid N-terminal Met-FLAG fusion protein of E. coli bacterial alkaline phosphatase (BAP) with a calculated molecular weight of 49.4 kDa. The Amino-terminal Met-FLAG-BAP control protein migrates as a 45-55 kDa band by SDS-PAGE depending on the electrophoretic conditions.

Other Notes

Control protein

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
FLAG-BAP is a trademark of Sigma-Aldrich Co. LLC


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library