Skip to Content
MilliporeSigma

Skip To

P5459

Protease from Bacillus licheniformis

glycerol solution (50%)

Synonym(s):

Proteinase from Bacillus licheniformis, Subtilisin A

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

CAS Number:
UNSPSC Code:
12352204
eCl@ss:
32160410
NACRES:
NA.54
EC Number:
MDL number:
Specific activity:
≥7 units/mg protein (biuret)
Biological source:
Bacillus licheniformis
Concentration:
≥50 mg/mL
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Bacillus licheniformis

Quality Segment

form

glycerol solution (50%)

specific activity

≥7 units/mg protein (biuret)

concentration

≥50 mg/mL

solubility

0.85% sodium chloride: soluble, clear (Diluted), H2O: miscible

storage temp.

2-8°C

General description

The enzyme hydrolyzes gelatin optimally at 50 °C over the range of pH 8-10. It is stable between pH 6-10 at 50 °C. Commercially, it is used in laundry detergent formulations and also for the recovery of silver from photographic emulsions.

Application

Protease from Bacillus licheniformis has been used to digest the neurolemma in the antennal lobe of the moth; Heliothis virescens.
Protease is an enzyme used to break down proteins by hydrolyzing peptide bonds. Protease is used to degrade proteins, to study protease inhibitors and to study thermal inactivation kinetics. Protease is used in nucleic acid isolation procedures in incubations. Protease from Bacillus licheniformis, product P5459, has been used to hydrolyze muscle during the isolation of mitochondria from skeletal muscle.

Biochem/physiol Actions

Protease catabolizes proteins by hydrolysis of peptide bonds. Proteases are inactivated by serine active-site inhibitors, such as phenylmethylsulfonyl fluoride (PMSF) and diisopropylfluorophosphate.

Packaging

Package size based on protein content.

Other Notes

One unit will hydrolyze casein to produce color equivalent to 1.0 μmole (181 μg) of tyrosine per min at pH 7.5 at 37 °C (color by Folin-Ciocalteu reagent), unless otherwise indicated.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P4860P5380P3910
description

glycerol solution (50%)

description

≥2.4 U/g

description

Type VIII, lyophilized powder, 7-15 units/mg solid

description

lyophilized powder, for use in Total Dietary Fiber Assay, TDF-100A

biological source

Bacillus licheniformis

biological source

-

biological source

Bacillus licheniformis

biological source

-

specific activity

≥7 units/mg protein (biuret)

specific activity

≥2.4 U/g

specific activity

7-15 units/mg solid

specific activity

-

form

glycerol solution (50%)

form

aqueous solution

form

lyophilized powder

form

lyophilized powder

concentration

≥50 mg/mL

concentration

-

concentration

-

concentration

-

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

−20°C

storage temp.

2-8°C

solubility

H2O: miscible, 0.85% sodium chloride: soluble, clear (Diluted)

solubility

-

solubility

10 mM NaAc (pH 7.5) and 5 mM CaAc: soluble, clear, H2O: soluble, colorless

solubility

-


Still not finding the right product?


Pictograms

Health hazardCorrosion

Signal Word

Danger

Hazard Codes

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Resp. Sens. 1

Storage Class

10 - Combustible liquids

WGK

WGK 2

PPE (personal protective equipment)

dust mask type N95 (US), Eyeshields, Faceshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters