Skip to Content
MilliporeSigma

P4912

Phospholipase D from Streptomyces sp.

Type VII, lyophilized powder, ≥150 units/mg solid

Synonym(s):

Lecithinase D, Phosphatidylcholine phosphatidohydrolase

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352204
NACRES:
NA.54
EC Number:
232-639-8
MDL number:
EC Number:
Specific activity:
≥150 units/mg solid
Biological source:
Streptomyces sp.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces sp.

Quality Segment

type

Type VII

form

lyophilized powder

specific activity

≥150 units/mg solid

storage temp.

−20°C

Application

Phospholipase D (PLD) has been used to hydrolyze the phosphate bonds of phospholipids and sphingomyelin to yield the corresponding phosphatidic acid. It has also been used to study metabolic labeling and direct imaging of choline phospholipids in vivo by measuring propargyl-Cho incorporation. Furthermore, PLD has been used in purification and kinetic studies.
The enzyme from Sigma has been used in the translocation of sphingosine kinase 1 (SK1) to membrane fractions under in vitro conditions. It has also been used to produce phosphatidic acid (PA) from phosphatidylcholine (PC) in HL60 permeabilized cells.

Biochem/physiol Actions

Phospholipase D is glycerophospholipid-specific. It is markedly less active on sphingomyelins and lysophospholipids. Phospholipase D hydrolyzes the phosphate bonds of phospholipids and sphingomyelin to give the corresponding phosphatidic acid.
Hydrolyzes the phosphate bonds of phospholipids and sphingomyelin to give the corresponding phosphatidic acid.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Analysis Note

Protein by biuret

Other Notes

One unit will liberate 1.0 μmole of choline from 1,2-Dioleoyl-sn-glycero-3-phosphocholine per hour at pH 8.0 at 37 °C.


pictograms

Health hazard

signalword

Danger

hcodes

Hazard Classifications

Resp. Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library