Skip to Content
MilliporeSigma

P4744

Phosphatase substrate

chromogenic, powder

Synonym(s):

4-Nitrophenyl phosphate disodium salt hexahydrate, pNPP disodium salt hexahydrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
O2NC6H4OP(O)(ONa)2 · 6H2O
CAS Number:
Molecular Weight:
371.14
NACRES:
NA.83
PubChem Substance ID:
UNSPSC Code:
12352204
EC Number:
224-246-5
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Phosphatase substrate, powder

Quality Segment

form

powder

solubility

water: 100 mg/mL, clear to slightly hazy

storage temp.

−20°C

SMILES string

O.O.O.O.O.O.[Na+].[Na+].[O-][N+](=O)c1ccc(OP([O-])([O-])=O)cc1

InChI

1S/C6H6NO6P.2Na.6H2O/c8-7(9)5-1-3-6(4-2-5)13-14(10,11)12;;;;;;;;/h1-4H,(H2,10,11,12);;;6*1H2/q;2*+1;;;;;;/p-2

InChI key

KAKKHKRHCKCAGH-UHFFFAOYSA-L

Application

Phosphatase substrate has been used as a substrate in tartrate-resistant acid phosphatase (TRAP) assay and alkaline phosphatase (ALP).
Chromogenic substrate for use with alkaline and acid phosphatase ELISA procedures. Produces a soluble end product that is yellow in color and can be read spectrophotometrically at 405 nm. The pNPP reaction may be stopped with 3 N NaOH and read at 405 nm.

Other Notes

Substrate for the determination of acid and alkaline phosphatases.


Still not finding the right product?

Explore all of our products under Phosphatase substrate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library