Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

P3804

Sigma-Aldrich

o-Phenylenediamine dihydrochloride

chromogenic, tablet

Synonym(s):

1,2-Phenylenediamine tablet, OPD, OPD Tablet

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(NH2)2·2HCl
CAS Number:
Molecular Weight:
181.06
Beilstein:
3912045
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.83

Product Name

o-Phenylenediamine dihydrochloride, tablet, 5 mg substrate per tablet

form

tablet

mp

258 °C (dec.) (lit.)

solubility

water: 1 tablet/10 mL, clear, colorless

storage temp.

2-8°C

SMILES string

Cl[H].Cl[H].Nc1ccccc1N

InChI

1S/C6H8N2.2ClH/c7-5-3-1-2-4-6(5)8;;/h1-4H,7-8H2;2*1H

InChI key

RIIWUGSYXOBDMC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

o-Phenylenediamine dihydrochloride (OPD) is a chromogenic substrate that can be used in enzyme-linked immunosorbent assay (ELISA) procedures as it utilizes horseradish peroxidase conjugates. The substrate produces a soluble end product that is orange-brown and can be read spectrophotometrically at 450 nm. The OPD reaction may be stopped with 3N HCl or 3M H2SO4 and read at 492 nm.

Application

o-Phenylenediamine dihydrochloride has been used:
  • as a chromogenic substrate in indirect enzyme-linked immunosorbent assay (ELISA) to detect immunoglobin G (IgG) antibodies against certain recombinant antigens/proteins
  • as a streptavidin-horseradish peroxidase (HRP) substrate sandwich ELISA to analyze human interleukin (IL)-17A, IL-23, and IP-10
  • as a substrate in ELISA for determination of IgG-class antibodies to Mycobacterium bovis in serum

Packaging

16 mg total tablet weight

Reconstitution

Dissolve one tablet in 0.05 M phosphate-citrate buffer, pH 5.0, to the desired concentration (typically an OPD concentration of 0.4 mg/ml is used). Add 40 μl of fresh 30% hydrogen peroxide per 100 ml of substrate buffer solution, immediately prior to use. Tablets are individually packaged in foil packets.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - Muta. 2 - Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Marco et al.
Peptides, 21(9), 1313-1321 (2000-11-10)
We have isolated a peptide from extracts of sinus glands from a South African spiny lobster species, Jasus lalandii, by high-performance liquid chromatography (HPLC) and identified it as a putative molt-inhibiting hormone (MIH) by (i) an in vitro assay with
Vilija Oke et al.
Arthritis research & therapy, 19(1), 139-139 (2017-06-18)
Interferon (IFN)-α is thought to have a pivotal role in systemic lupus erythematosus (SLE), and type III IFNs (IFN-λ) were recently also associated with SLE. In this study, we measured levels of IFN-α, IFN-λ1, and related cytokines, such as IL-17A
Joanne L Casey et al.
Methods in molecular biology (Clifton, N.J.), 421, 111-124 (2008-10-02)
Large repertoires of peptides displayed on bacteriophage have been extensively used to select for ligand-binding molecules. This is a relatively straightforward process involving several cycles of selection against target molecules, and the resulting ligands can be tailored to various applications.
Katherine A Deets et al.
eLife, 10 (2021-12-24)
The innate immune system detects pathogens and initiates adaptive immune responses. Inflammasomes are central components of the innate immune system, but whether inflammasomes provide sufficient signals to activate adaptive immunity is unclear. In intestinal epithelial cells (IECs), inflammasomes activate a
Zahra Hemati et al.
PloS one, 15(6), e0233695-e0233695 (2020-06-02)
Johne's disease (JD) is an infectious wasting condition of ruminants caused by Mycobacterium avium subsp. paratuberculosis (MAP) in domestic livestock of every country that has been investigated. Controlling JD is problematic due to the lack of sensitive, specific, efficient, and

Articles

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

NBT-BCIP substrate system aids in western blotting and immunohistological staining, producing a blue-purple insoluble end product.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service