Skip to Content
MilliporeSigma

Skip To

P3657

Pyridoxal 5′-phosphate hydrate

powder, BioReagent, suitable for cell culture

Synonym(s):

3-Hydroxy-2-methyl-5-([phosphonooxy]methyl)-4-pyridinecarboxaldehyde, Codecarboxylase, PLP, Pyridoxal 5-phosphate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H10NO6P · xH2O
CAS Number:
Molecular Weight:
247.14 (anhydrous basis)
UNSPSC Code:
12352205
NACRES:
NA.75
PubChem Substance ID:
EC Number:
200-208-3
Beilstein/REAXYS Number:
234749
MDL number:
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


product line

BioReagent

Quality Segment

assay

≥98%

form

powder

mol wt

Mw 247.14 g/mol

technique(s)

cell culture | mammalian: suitable

color

off-white to yellow-brown

mp

140-143 °C

solubility

1 M HCl: soluble 50 mg/mL, clear, colorless to light yellow-green

storage temp.

−20°C

SMILES string

CC1=NC=C(COP(O)(O)=O)C(C([H])=O)=C1O

InChI

1S/C8H10NO6P/c1-5-8(11)7(3-10)6(2-9-5)4-15-16(12,13)14/h2-3,11H,4H2,1H3,(H2,12,13,14)

InChI key

NGVDGCNFYWLIFO-UHFFFAOYSA-N

General description

Pyridoxal-phosphate (PLP, pyridoxal-5′-phosphate, P5P) is a prosthetic group of some enzymes. It is the active form of vitamin B6, which comprises three natural organic compounds, pyridoxal, pyridoxamine and pyridoxine. PLP acts as a coenzyme in all transamination reactions, and in some decarboxylation and deamination reactions of amino acids.

Application


  • Hierarchical Surface Architecture of Hemodialysis Membranes for Eliminating Homocysteine Based on the Multifunctional Role of Pyridoxal 5′-phosphate.: Discusses the development of advanced hemodialysis membranes using pyridoxal 5′-phosphate, highlighting its role in reducing homocysteine levels, which could contribute to improved outcomes for dialysis patients (Jiang P et al., 2020).


Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
P925582870P6155
description

powder, BioReagent, suitable for cell culture

description

≥98%

description

≥97.0% (NT)

description

BioReagent, suitable for cell culture, suitable for insect cell culture

technique(s)

cell culture | mammalian: suitable

technique(s)

HPLC: suitable

technique(s)

enzyme immunoassay: suitable

technique(s)

cell culture | mammalian: suitable, cell culture | insect: suitable

assay

≥98%

assay

≥98%

assay

≥97.0% (NT)

assay

≥99% (HPLC)

form

powder

form

powder

form

powder

form

powder

storage temp.

−20°C

storage temp.

−20°C

storage temp.

2-8°C

storage temp.

−20°C

solubility

1 M HCl: soluble 50 mg/mL, clear, colorless to light yellow-green

solubility

-

solubility

-

solubility

water: soluble 50 mg/mL, clear, colorless to light greenish-yellow

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200


Still not finding the right product?

Explore all of our products under Pyridoxal 5′-phosphate hydrate


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable

PPE (personal protective equipment)

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library




Questions

Reviews

No rating value

Active Filters