Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
Empirical Formula (Hill Notation):
C23H33NO4
CAS Number:
Molecular Weight:
387.51
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.25
MDL number:
Form:
powder
Assay:
≥98.00% (TLC)
Product Name
Progesterone 3-(O-carboxymethyl)oxime,
assay
≥98.00% (TLC)
Quality Segment
form
powder
technique(s)
ELISA: suitable
solubility
chloroform: 49-51 mg/mL, clear, colorless to faintly yellow
shipped in
ambient
storage temp.
room temp
SMILES string
[H][C@@]12CCC3=CC(\CC[C@]3(C)[C@@]1([H])CC[C@]4(C)[C@H](CC[C@@]24[H])C(C)=O)=N\OCC(O)=O
InChI
1S/C23H33NO4/c1-14(25)18-6-7-19-17-5-4-15-12-16(24-28-13-21(26)27)8-10-22(15,2)20(17)9-11-23(18,19)3/h12,17-20H,4-11,13H2,1-3H3,(H,26,27)/b24-16+/t17-,18+,19-,20-,22-,23+/m0/s1
InChI key
PPELYUTTZHLIAZ-CDUDAXDSSA-N
Application
Progesterone 3-(O-carboxymethyl) oxime can be used in microtitre plate enzyme immunoassays (EIAs) for progesterone.
Biochem/physiol Actions
Progesterone 3-(O-carboxymethyl) oxime can be used for studying the steroid specificity of progestin (Pi) binding. The result shows that this product has a reasonable degree of specificity for binding to Pi receptor in the thymus.
Preparation Note
Progesterone 3-(O-carboxymethyl) oxime dissolves in chloroform at 50 mg/ml to yield a clear, colorless to faint yellow solution.
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
