Skip to Content
MilliporeSigma

P3190

TWEEN® 65

Synonym(s):

Polyoxyethylenesorbitan Tristearate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12161900
NACRES:
NA.25
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

non-ionic

Quality Segment

form

solid

composition

Stearic acid, ≥40% (balance primarily palmitic acid)

concentration

≥35% (palmitic acid, GC)

technique(s)

DNA extraction: suitable, RNA extraction: suitable, cell culture | mammalian: suitable, protein purification: suitable

General description

TWEEN 65 is a fatty acid polyoxyethylene ester of sorbitan.

Application

TWEEN 65 has been used in a study as a carbon source for the reductive dechlorination of hexachlorobenzene in a mixed, methanogenic culture. It has also been used in a study to investigate its ability to stimulate growth of the hypocotyl of Amaranthus caudatus L.

Legal Information

TWEEN is a registered trademark of Croda International PLC


Storage Class

10 - Combustible liquids

wgk

WGK 1

flash_point_f

300.2 °F - closed cup

flash_point_c

149.00 °C - closed cup

ppe

Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library