Skip to Content
MilliporeSigma

P2658

Poly-L-lysine hydrochloride

suitable for cell culture, Mol wt 15,000-30,000

Synonym(s):

L-Lysine hydrochloride homopolymer

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352209
eCl@ss:
32160406
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Poly-L-lysine hydrochloride, mol wt 15,000-30,000

form

powder or solid

Quality Segment

mol wt

15,000-30,000

technique(s)

cell culture | mammalian: suitable

color

white to light yellow

application(s)

cell analysis

storage temp.

−20°C

Application

Poly-lysine with mol. wt. >70,000 is useful in promoting cell adhesion to solid substrates.
Poly-L-lysine hydrochloride has been used to coat glass slides and coverslips to facilitate cell attachment.

Biochem/physiol Actions

L-Lysine is believed to prevent osteoporosis of bones, which is a characteristics of vascular calcification. Glutaric aciduria type I and pyridoxine-dependent epilepsy occurs due to lack of L-lysine catabolic pathway. Lysine participates in the regulation of NO (nitric oxide) synthesis. It possesses antiviral activity, therefore it is used in the treatment of Herpes simplex. L-Lysine is associated with a number of biochemical reactions such as acetylation, O-linked glycosylation, ubiquitination and protein methylation. Poly-L-lysine is considered as a biocompatible polymer and serves for the purpose of drug delivery across cell membrane. It is useful as one of the nanosized contrast agents in MRI (magnetic resonance imaging).

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.


Still not finding the right product?

Explore all of our products under Poly-L-lysine hydrochloride


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library