Skip to Content
MilliporeSigma

P2534

Poly-L-histidine hydrochloride

mol wt ≥5000

Synonym(s):

L-Histidine homopolymer hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


form

powder

Quality Segment

mol wt

≥5000

color

white to very light yellow

storage temp.

−20°C

SMILES string

N[C@H](Cc1c[nH]cn1)C(O)=O

InChI key

HNDVDQJCIGZPNO-RXMQYKEDSA-N

Analysis Note

Molecular weight by MALLS.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library