Skip to Content
MilliporeSigma

P2363

Protein Kinase G II from rat

>90% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf9 cells, solution

Synonym(s):

Protein Kinase, 3′,5′-Cyclic GMP Dependent from rat

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

UNSPSC Code:
41106305
NACRES:
NA.32
MDL number:
Specific activity:
~0.7 U/mg
Assay:
>90% (SDS-PAGE)
Recombinant:
expressed in baculovirus infected Sf9 cells
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


recombinant

expressed in baculovirus infected Sf9 cells

Quality Segment

assay

>90% (SDS-PAGE)

form

solution

specific activity

~0.7 U/mg

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

Gene Information

rat ... Prkg2(25523)

General description

Protein kinase, cGMP-dependent, type II (PRKG2), also known as type II cGMP-dependent protein kinase (cGK), is encoded by the gene mapped to human chromosome 4q13.1–q21.1.

Biochem/physiol Actions

Protein kinase, cGMP-dependent, type II (PRKG2) is implicated in the regulation of intestinal fluid balance in man. The encoded protein plays an essential role in proliferative to hypertrophic transition of growth plate chondrocytes during endochondral ossification. Loss of PRKG2 function is associated with the development of renal cysts, intellectual disability and speech defect. In rodent and bovine models, deletion of the gene leads to dwarfism.

Other Notes

One unit will phosphorylate one μmole of VASPtide (RRKVSKQE) per minute at pH 7.4


Storage Class

10 - Combustible liquids

wgk

WGK 2

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library