Skip to Content
MilliporeSigma
All Photos(1)

Documents

P1243

Sigma-Aldrich

pT7-FLAG-2 Expression Vector

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106610
NACRES:
NA.85

recombinant

expressed in E. coli

grade

for molecular biology

shipped in

dry ice

storage temp.

−20°C

General description

pT7-FLAG-2 is a 4.8 kb E . coli expression vector used for cloning and cytoplasmic expression of a properly inserted open reading frame as a C-terminal FLAG® fusion protein containing the FLAG epitope (DYKDDDDK). The promoter region of the very strong phage T7 promoter drives transcription of ORFFLAG fusion constructs. This vector requires the use of E. coli cells containing a source of the T7 polymerase, such as BL21(DE3)T1R cells, Catalog Number B2935.

The C-terminal FLAG fusion protein may be detected using Monoclonal ANTI-FLAG?M2, Catalog Number
F3165, and purified using the ANTI-FLAG M2 Affinity Gel, Catalog Number A2220.

Vector Maps and Sequences

Components

  • pT7-FLAG-2 Expression Vector, 10 μg (P6867) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.
  • pT7-FLAG-2-BAP Control Vector 1 μg ( P7117) is supplied as 0.5 mg/ml in 10 mM Tris-HCl (pH 8.0) with 1 mM EDTA.

Legal Information

FLAG is a registered trademark of Merck KGaA, Darmstadt, Germany
pT7-FLAG is a trademark of Sigma-Aldrich Co. LLC

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jessica Radzio et al.
Retrovirology, 8, 69-69 (2011-08-24)
N348I in HIV-1 reverse transcriptase (RT) confers resistance to zidovudine (AZT) and nevirapine. Biochemical studies demonstrated that N348I indirectly increases AZT resistance by decreasing the frequency of secondary ribonuclease H (RNase H) cleavages that reduce the RNA/DNA duplex length of
Seung-Gi Jin et al.
The Journal of biological chemistry, 280(13), 12700-12709 (2005-02-11)
MBD2 and MBD3 are two proteins that contain methyl-CpG binding domains and have a transcriptional repression function. Both proteins are components of a large CpG-methylated DNA binding complex named MeCP1, which consists of the nucleosome remodeling and histone deacetylase complex
Lingyan Wang et al.
Communications biology, 4(1), 921-921 (2021-07-31)
Retinoic acid-inducible gene I (RIG-I) senses viral RNA and instigates an innate immune signaling cascade to induce type I interferon expression. Currently, the regulatory mechanisms controlling RIG-I activation remain to be fully elucidated. Here we show that the FAK family

Articles

Bacterial Expression Vectors: T7 Promoter System. T7 Vectors for Highest Expression Levels in Bacteria.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service