Skip to Content
MilliporeSigma

P0184

Pseudomonas exotoxin A

from Pseudomonas aeruginosa, protein synthesis inhibitor, lyophilized powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

CAS Number:
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Form:
lyophilized powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Pseudomonas exotoxin A from Pseudomonas aeruginosa, lyophilized powder

form

lyophilized powder

Quality Segment

composition

Protein, ~30% E1%/280

storage temp.

2-8°C

General description

Pseudomonas exotoxin A is secreted by Pseudomonas aeruginosa. It is a member of the family mono-ADP-ribosyltransferases.

Biochem/physiol Actions

Pseudomonas exotoxin A functions as an anticancer agent.
It catalyses the transfer of ADP-ribose moiety of NAD+ to the histidine imidazole ring of diphthamide.
Shown to be toxic to animals and to cell lines, and to inhibit protein synthesis via ADP ribosylation of elongation factor 2.

Packaging

Package size based on protein content.

Physical form

Lyophilized powder containing Tris buffer, EDTA, NaCl and lactose


pictograms

Skull and crossbones

signalword

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library