Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NA.47
UNSPSC Code:
12171500
Quality Level
color
colorless
density
1.030 g/cm3
application(s)
hematology
histology
storage temp.
room temp
General description
PolyFreeze is a water-soluble support matrix and a form of embedding medium for frozen sectioning. PolyFreeze sections flawlessly at routine frozen specimen thickness of 5-6 μ and leaves no slide residue. It provides for -20 ºC cryostat sectioning in rapid set embedment. This medium freezes quickly supporting the tissue for sectioning at 3 mm and up with no cracking of the matrix at temperatures from -8 °C to -25 °C. Snap freeze using PolyFreeze with isopentane and liquid nitrogen, or dry ice (slush/slurry or bunker). Store specimens frozen in PolyFreeze in liquid nitrogen canisters or in airtight containers in a -80 °C freezer. PolyFreeze does not cause autofluorescence and can easily be washed away during fixation and rinsed prior to staining. Once rinsed, there is no trace of the support matrix to interfere with staining or immunohistochemistry (IHC) reactions.
Application
PolyFreeze has been used as an embedding medium for frozen sectioning of plant and animal samples.
Preparation Note
PolyFreeze is supplied ready-to-use. A cloudy appearance of the unfrozen product has no effect on its performance.
Store the product at room temperature
Storage Class
10 - Combustible liquids
wgk
WGK 1
flash_point_f
Not applicable
flash_point_c
Not applicable
ppe
Eyeshields, Gloves, multi-purpose combination respirator cartridge (US)
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Related Content
Instructions
Application of Endophytic Pseudomonas fluorescens and a Bacterial Consortium to Brassica napus Can Increase Plant Height and Biomass under Greenhouse and Field Conditions.
Lally R D, et al.
Frontiers in Plant Science, 8, 2193-2193 (2017)
Gamze Turgay-İzzetoğlu et al.
Acta histochemica, 121(4), 460-471 (2019-04-14)
The fat body, originates from mesoderm, has many metabolic functions which changes as the embryonic development of the insect progresses. It plays an important role in the intermediate metabolism and in the metabolism of proteins, lipids and carbohydrates. It has
Identification of a Potential Common Ancestor for Mammalian Cross-Presenting Dendritic Cells in Teleost Respiratory Surfaces.
Soleto I, et al.
Frontiers in Immunology, 9, 59-59 (2018)
Global Trade Item Number
| SKU | GTIN |
|---|---|
| P0091-6X120ML | 04061834290780 |