OH30429846
Ohaus® Pioneer® Analytical Balance
Model PX124/E, weighing capacity 120 g, resolution: 0.0001 g, AC/DC input 110 V, US 3-pin plug (North America)
Synonym(s):
high accuracy weighing balances, high precision balance, high precision weighing, laboratory balances
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41111502
Recommended Products
material
metal weighing plate
feature
CSA approved
packaging
pack of 1 ea
manufacturer/tradename
Ohaus® 30429846
weighing capacity measuring range
120 g, resolution: 0.0001 g
weighing pan diam.
9 cm
Looking for similar products? Visit Product Comparison Guide
General description
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
Application
Basic Weighing, Parts Counting, Percent Weighing, Animal Weighing, Density Determination
Features and Benefits
The Pioneer PX combines essential weighing functionality with competitive performance, offering high accuracy and repeatability for applications in laboratory, industrial and education settings. The PX is affordably priced, and intuitively designed for intelligent operation with a second line display for additional information, and USB and RS232 connectivity for easy communication.
Legal Information
Ohaus is a registered trademark of Ohaus Corp.
Pioneer is a registered trademark of Ohaus Corp.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service