Skip to Content
MilliporeSigma

O7639

Org 24598 lithium salt

≥98% (HPLC), solid

Synonym(s):

R-(−)-N-Methyl-N-[3-[(4-trifluoromethyl)phenoxy]-3-phenyl-propyl]glycine lithium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C19H19F3LiNO3
CAS Number:
Molecular Weight:
373.30
UNSPSC Code:
12352202
PubChem Substance ID:
MDL number:
Assay:
≥98% (HPLC)
Form:
solid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

solid

color

white to off-white

solubility

H2O: >2 mg/mL

storage temp.

2-8°C

SMILES string

[Li+].CN(CC[C@@H](Oc1ccc(cc1)C(F)(F)F)c2ccccc2)CC([O-])=O

InChI

1S/C19H20F3NO3.Li/c1-23(13-18(24)25)12-11-17(14-5-3-2-4-6-14)26-16-9-7-15(8-10-16)19(20,21)22;/h2-10,17H,11-13H2,1H3,(H,24,25);/q;+1/p-1/t17-;/m1./s1

InChI key

VMQXVSNARQMSDL-UNTBIKODSA-M

Biochem/physiol Actions

Org 24598 is a selective, potent inhibitor of glial GlyT (GlyT1, glycine transporter type 1).
Org 24598 is a selective, potent inhibitor of glial GlyT (GlyT1, glycine transporter type 1). In rats (P12-P16) and in the presence of kynurenic acid, 6-cyano-7-nitroquinoxaline-2,3-dione (CNQX) and bicuculline, ORG 24598 at a concentration of 10 μM induced a mean inward current of -10/-50 pA at -60 mV and increased significantly the decay time constants of miniature (mIPSCs), spontaneous (sIPSCs) and electrically evoked glycinergic (eIPSCs) inhibitory postsynaptic currents. Replacing extracellular sodium with N-methyl-d-glucamine or superfusing the slice with micromolar concentrations of glycine also increased the decay time constant of glycinergic IPSCs. Glycine (1-5 μM and d-serine (10 μM) increased the amplitude of eEPSCs whereas l-serine had no effect. Org 24598 increased significantly the amplitude of NMDA receptor-mediated eEPSCs without affecting the amplitude of non-NMDA receptor-mediated eEPSCs. This brings conclusion that blocking glial glycine transporter by Org 24598 increased the level of glycine in spinal cord slices, which in turn prolonged the duration of glycinergic synaptic current and potentiated the NMDA-mediated synaptic response.

Features and Benefits

This compound is featured on the Glycine Transporters page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library