Skip to Content
MilliporeSigma

N8501

5-Nitro-1,10-phenanthroline

≥97%, crystalline

Synonym(s):

Nitroferroin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C12H7N3O2
CAS Number:
Molecular Weight:
225.20
UNSPSC Code:
12352202
NACRES:
NA.77
PubChem Substance ID:
EC Number:
224-097-6
Beilstein/REAXYS Number:
196245
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

synthetic

Quality Segment

assay

≥97%

form

crystalline

color

yellow to orange

mp

202-204 °C (lit.)

solubility

ethanol: 50 mg/mL, clear to slightly hazy, yellow to orange

SMILES string

[O-][N+](=O)c1cc2cccnc2c3ncccc13

InChI

1S/C12H7N3O2/c16-15(17)10-7-8-3-1-5-13-11(8)12-9(10)4-2-6-14-12/h1-7H

InChI key

PDDBTWXLNJNICS-UHFFFAOYSA-N

Biochem/physiol Actions

Platinum chelates of a variety of 5-substituted phenanthrolines, including this compound, were tested for cytotoxicity against L1210 murine leukemia cells. Though all tested compounds had very similar DNA-binding affinities, they exhibited a wide range of cytotoxicity.


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library