Skip to Content
MilliporeSigma

N4764

2-Nitrophenyl β-D-cellobioside

≥98% (TLC), powder

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(β-D-Glc-[1→4]-β-D-Glc-1→OC6H4NO
CAS Number:
Molecular Weight:
463.39
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

2-Nitrophenyl β-D-cellobioside,

assay

≥98% (TLC)

Quality Segment

form

powder

solubility

water: 50 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

OCC1OC(OC2C(O)C(O)C(OC2CO)Oc3ccccc3N(=O)=O)C(O)C(O)C1O

InChI

1S/C18H25NO13/c20-5-9-11(22)12(23)14(25)18(30-9)32-16-10(6-21)31-17(15(26)13(16)24)29-8-4-2-1-3-7(8)19(27)28/h1-4,9-18,20-26H,5-6H2

InChI key

CYCLRDYAFVRUDE-UHFFFAOYSA-N



Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library