Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N4663

Sigma-Aldrich

Anti-Nurr1 antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NOT, Anti-NR4A2, Anti-Nuclear receptor of T cells, Anti-TINUR

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

concentration

~1 mg/mL

technique(s)

immunohistochemistry: 1:100-1:200
western blot: 1:1,000-1:2,000

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NR4A2(4929)
mouse ... Pls3(18827)
rat ... Nr4a2(54278)

General description

Nur-related factor 1 (Nurr1) belongs to nuclear receptor super family. It is a brain-specific transcription factor, confined to central nervous system. Additionally, Nurr1 also modulates gene expression by repressing other transcriptional factors. Nurr1 is expressed in developing and mature dopamine neurons, hippocampus and cerebral cortex. It is crucial for the early development of dopamine neurons in midbrain during development, protection and survival of the neurons. Nurr1 may play a very critical role in the development of Parkinson′s disease and other neurodegenerative diseases and in human atherosclerotic lesions
Anti- Nurr1 recognizes human Nurr1.

Immunogen

synthetic peptide corresponding to an internal sequence of human Nurr1.

Application

Anti-Nurr 1 antibody may be used in immunoblotting at a working dilution of 1:1000 to 1:2000. For immunohistochemistry the recommended dilution is 1:100 to 1:200.

Physical form

Solution in phosphate buffered saline, containing 0.02% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asa Wallen-Mackenzie et al.
Genes & development, 17(24), 3036-3047 (2003-12-19)
The retinoid X receptor (RXR) is essential as a common heterodimerization partner of several nuclear receptors (NRs). However, its function as a bona fide receptor for endogenous ligands has remained poorly understood. Such a role would depend on the existence
Peter I Bonta et al.
Circulation, 121(18), 2023-2032 (2010-04-28)
Restenosis is the major drawback of percutaneous coronary interventions involving excessive activation and proliferation of vascular smooth muscle cells (SMCs). The nuclear receptor Nurr1 is an early response gene known mainly for its critical role in the development of dopamine
Banafsheh Kadkhodaei et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 29(50), 15923-15932 (2009-12-18)
Transcription factors involved in the specification and differentiation of neurons often continue to be expressed in the adult brain, but remarkably little is known about their late functions. Nurr1, one such transcription factor, is essential for early differentiation of midbrain
S W Law et al.
Molecular endocrinology (Baltimore, Md.), 6(12), 2129-2135 (1992-12-01)
We have identified and cloned a novel member of the nuclear receptor superfamily. The cDNA was isolated from a mouse brain cDNA library and encodes a protein 598 amino acids in length with a predicted mol wt of 66 kilodaltons.
Yinxiu Ding et al.
Molecular medicine reports, 14(3), 1993-1999 (2016-07-20)
Parkinson's disease (PD) is a severe neurodegenerative disorder. Although the detailed underlying molecular mechanism remains to be elucidated, the major pathological feature of PD is the loss of dopaminergic (DA) neurons of the substantia nigra. The use of donor stem

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service