Skip to Content
MilliporeSigma

N115

Naltrindole hydrochloride

≥97.5% (HPLC), solid, δ-opioid receptor antagonist

Synonym(s):

NTI hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C26H26N2O3 · HCl
CAS Number:
Molecular Weight:
450.96
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
MDL number:
Form:
solid
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Naltrindole hydrochloride, solid

form

solid

Quality Segment

storage condition

desiccated

color

off-white

solubility

methanol: 4.5 mg/mL, H2O: 8 mg/mL

storage temp.

−20°C

SMILES string

Cl[H].Oc1ccc2C[C@H]3N(CC[C@@]45[C@@H](Oc1c24)c6[nH]c7ccccc7c6C[C@@]35O)CC8CC8

InChI

1S/C26H26N2O3.ClH/c29-19-8-7-15-11-20-26(30)12-17-16-3-1-2-4-18(16)27-22(17)24-25(26,21(15)23(19)31-24)9-10-28(20)13-14-5-6-14;/h1-4,7-8,14,20,24,27,29-30H,5-6,9-13H2;1H/t20-,24+,25+,26-;/m1./s1

InChI key

KNJKRQXCFJCQHC-IDRHMUJXSA-N

Gene Information

human ... OPRD1(4985)

Biochem/physiol Actions

Highly selective, non-peptide δ-opioid receptor antagonist that crosses the blood-brain barrier.

Features and Benefits

This compound is featured on the Opioid Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Gloves, type N95 (US)



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library