Skip to Content
MilliporeSigma
All Photos(3)

Key Documents

N1143

Sigma-Aldrich

1-(4-Nitrophenyl)glycerol

crystalline

Synonym(s):

α-p-Nitrophenylglycerol, 1-(4-Nitrophenyl)-1,2,3-propanetriol, PNPG

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
O2NC6H4CH(OH)CH(OH)CH2OH
CAS Number:
Molecular Weight:
213.19
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

crystalline

Quality Level

color

off-white

mp

98-101 °C (lit.)

storage temp.

2-8°C

SMILES string

OCC(O)C(O)c1ccc(cc1)[N+]([O-])=O

InChI

1S/C9H11NO5/c11-5-8(12)9(13)6-1-3-7(4-2-6)10(14)15/h1-4,8-9,11-13H,5H2

InChI key

IUZVZBIQZKBWCC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-(4-Nitrophenyl)glycerol may be used as a linear small saccharide model in studies involving the molecular recognition mechanism between a synthetic glycophane receptor and carbohydrates in water.
Useful additive for controlling swarming of Proteus bacteria on culture media.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Molecular recognition of carbohydrates using a synthetic receptor. A model system to understand the stereoselectivity of a carbohydrate-carbohydrate interaction in water.
Jimenez-Barbero J
Journal of the American Chemical Society, 117(45), 11198-11204 (1995)
Williams, F.
Abstracts Society for Microbiology (1969)
R Kopp et al.
Applied microbiology, 14(6), 873-878 (1966-11-01)
The effects of sodium tetradecyl sulfate (STS), beta-phenethyl alcohol (PEA), and p-nitrophenylglycerol (PNPG) on motility, swarming, flagellation, and growth of Proteus were examined. Growth-inhibitory concentrations (GIC) and swarming-inhibitory concentrations (SIC) were determined. A characterization of the swarming-inhibitory efficacy of these

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service