Skip to Content
MilliporeSigma

N1019

(−)-Nicotine hemisulfate salt

≥95% (TLC), ~35 % (w/v) in H2O (based on free base)

Synonym(s):

(−)-1-Methyl-2-(3-pyridyl)pyrrolidine

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Linear Formula:
(C10H14N2)2 · H2SO4
CAS Number:
Molecular Weight:
422.54
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352210
MDL number:
Assay:
≥95% (TLC)
Form:
liquid
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (TLC)

form

liquid

concentration

~35 % (w/v) in H2O (based on free base)

color

dark brown

SMILES string

OS(O)(=O)=O.CN1CCC[C@H]1c2cccnc2.CN3CCC[C@H]3c4cccnc4

InChI

1S/2C10H14N2.H2O4S/c2*1-12-7-3-5-10(12)9-4-2-6-11-8-9;1-5(2,3)4/h2*2,4,6,8,10H,3,5,7H2,1H3;(H2,1,2,3,4)/t2*10-;/m00./s1

InChI key

IECQULMJVNSKDB-RCWTXCDDSA-N

Gene Information

General description

Nicotine is found in the leaves of Nicotiana species. It binds to nicotinic acetylcholine receptors and inhibits endogenous neurotransmitters. Nicotine is associated with lung cancer development and reduces the efficiency of chemotherapeutic agents. It is used to treat Alzheimer′s and Parkinson′s disease.

Application

(−)-Nicotine hemisulfate salt has been used to study the influence of nicotine on the severity of choroidal neovascularization (CNV). It also has been used as a natural nectar alkaloid to study the effect of self-medication in bumblebees.

Biochem/physiol Actions

Prototype nicotinic acetylcholine receptor agonist; naturally occurring isomer.

Analysis Note

Mixture of nicotine base and nicotine sulfate.

Disclaimer

Shelf-life of the solution as supplied is at least six years when stored at room temperature.


pictograms

Skull and crossbonesEnvironment

signalword

Danger

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 2 Inhalation - Acute Tox. 3 Oral - Aquatic Chronic 2

Storage Class

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library