Skip to Content
MilliporeSigma

N0665

NDSB 211

≥98% (TLC)

Synonym(s):

3-[Dimethyl-(2-hydroxyethyl)ammonio]-1-propanesulfonate, Dimethyl(2-hydroxyethyl)(3-sulfopropyl)ammonium hydroxide inner salt, Dimethyl-2-hydroxyethylammoniumpropane sulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H17NO4S
CAS Number:
Molecular Weight:
211.28
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12161900
EC Number:
254-176-0
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


description

zwitterionic

Quality Segment

assay

≥98% (TLC)

form

solid

mol wt

211.28 g/mol

technique(s)

DNA extraction: suitable

color

white

solubility

methanol: 5 mg/mL, clear, colorless

SMILES string

OCC[N+](C)(C)CCCS([O-])(=O)=O

InChI

1S/C7H17NO4S/c1-8(2,5-6-9)4-3-7-13(10,11)12/h9H,3-7H2,1-2H3

InChI key

CNXPCGBLGHKAIL-UHFFFAOYSA-N

General description

Dimethyl-2-hydroxyethylammoniumpropane sulfonate, or NDSB 211, is a nondetergent-sulfobetain. It is zwitterionic over a wide pH range, easily removed by dialysis and shows no significant absorption in the near UV range.

Application

Non-detergent sulfobetaine is a compound used for non-denaturing protien purification. Increases the extraction yield of membrane, nuclear and cytoskeletal associated proteins. Zwitterionic over a wide pH range, easily removed by dialysis and no significant absorption in the near UV range. Specific applications include microsomal protein extraction, nuclear protein recovery, precipitation reduction in IEF, and membrane-bound protease purification.
NDSB 211 has been used in a study to assess halophilic protein stabilization by the mild solubilizing agents nondetergent sulfobetaines. It has also been used in a study to investigate an alternative strategy for crystallizing macromolecules.

Other Notes

This product is non-micelle forming.


pictograms

Corrosion

signalword

Danger

hcodes

Hazard Classifications

Skin Corr. 1B

Storage Class

8A - Combustible corrosive hazardous materials

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library