Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

N0649

Sigma-Aldrich

Anti-Neuronal Specific Enolase antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-NSE

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, rat, human

technique(s)

immunohistochemistry: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

2-8°C

target post-translational modification

unmodified

Gene Information

human ... ENO2(2026)

General description

Neuronal specific enolase 2 (ENO2) is also known as γ-enolase. It is present in the cytoplasm of neuroendocrine cells.

Immunogen

brain NSE from human.

Biochem/physiol Actions

Neuronal specific enolase 2 (ENO2) has been studied as a biomarker in the progression of multiple sclerosis. It converts 2-phosphoglycerate into phosphoenolpyruvate.

Physical form

Prediluted in 20 mM HEPES, containing 135 mM NaCI, 1% BSA, and 0.09% sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcus W Koch et al.
Journal of the neurological sciences, 350(1-2), 46-50 (2015-02-18)
Previous studies suggested that serum neuron specific enolase (NSE) may be a biomarker associated with progression in MS. We measured serum NSE levels in 385 patients with multiple sclerosis (MS) (264 with relapsing-remitting (RR) MS, 86 with secondary progressive (SP)
Jianbo Li et al.
Diabetes care, 36(11), 3405-3410 (2013-07-13)
To explore the relationship between serum neuron-specific enolase (NSE) levels and diabetic neuropathy. Type 1 or 2 diabetic and healthy control subjects (n = 568) were randomly enrolled in a cross-sectional study. Diabetic neuropathy status was documented by the presence
Yoshinori Kawai
Frontiers in neuroanatomy, 12, 63-63 (2018-08-09)
To integrate and broadcast neural information, local microcircuits and global macrocircuits interact within certain specific nuclei of the central nervous system. The structural and functional architecture of this interaction was determined for the caudal nucleus of the tractus solitarius (NTS)
Shu-Lin Guo et al.
Developmental neuroscience, 44(2), 91-101 (2022-01-06)
Stem cell-based therapy has been evaluated in many different clinical trials for various diseases. This capability was applied in various neurodegenerative diseases, such as multiple sclerosis, which is characterized by demyelination, axonal injury, and neuronal loss. Dental pulp stem cells

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service