Key Documents
MTCC3175AST
MTC™ Bio ClearSeal™ Microtubes screw caps
capacity × W 2.0 mL, non-sterile, assorted colors cap, self standing, pack of 1000 ea
Synonym(s):
screw-cap, micro, centrifuge, tube, conical
About This Item
UNSPSC Code:
41122500
NACRES:
NB.11
material
assorted colors cap
polypropylene
round
screw top cap
sterility
non-sterile
packaging
pack of 1000 ea
manufacturer/tradename
MTC Bio C3175-AST
capacity × W
2.0 mL
General description
Screw cap for 2.0mL microtube MTC3172-1000EA, assorted colors (red, blue, green, yellow, orange), non-sterile, with installed O-ring. Packaged as 5 Individually colored bags of 200 caps, total 1000 caps.
Features and Benefits
  • Ultra-clear polypropylene
  • Non-graduated
  • Resealable bags of 50
  • Leak-proof O-ring seal from -86°C to +121°C
  • Sterile, RNase, DNase, Pyrogen free
Legal Information
ClearSeal is a trademark of MTC Bio
MTC is a trademark of MTC Bio
Choose from one of the most recent versions:
Certificates of Analysis (COA)