Key Documents
MTCB1204
MTC Bio MultiBox for Western Blotting
4 compartments, L × W × D 38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.), for various gels including half-mini protein gels
Synonym(s):
gel processing box, multi gel processing box
About This Item
UNSPSC Code:
41121800
NACRES:
NB.15
material
polystyrene
packaging
pack of 6 ea
manufacturer/tradename
MTC Bio B1204
L × W × D
38 mm (1 1/2 in.) × 71 mm (2 13/16 in.) × 28 mm (1 1/8 in.)
General description
MultiBox, 4 compartments, 38 x 71 x 28mm each (11/2 x 2 13/16 x 11/8 in.), for various gels including half-mini protein gels, 6/pk
The MultiBox Western Blotting Box provides the ease and convenience of using one box to process multiple gels, allowing users to compare results side by side in real time.
Features and Benefits
  • A perfect box for every blot
  • Reduce antibody usage
  • Hinged lid minimizes evaporation
  • For halves, wholes, strips or minis
Legal Information
MTC is a trademark of MTC Bio
MultiBox is a trademark of MTC Bio