MilliporeSigma
MTCA9000C
MTC Bio Autoclave Bags
W × H 12 in. (31 cm) × 26 in. (66 cm), clear (blue printed warnings)
Synonym(s):
biohazard bag, Autoclave pouches, MTC Bio A9000C, Steam sterilization bags, sterilization bag
Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
NACRES:
NB.15
UNSPSC Code:
24111500
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist
material
clear (blue printed warnings), polypropylene
packaging
pack of 200 ea (bulk bag)
manufacturer/tradename
MTC Bio A9000C
W × H
12 in. (31 cm) × 26 in. (66 cm)
General description
Autoclave bags, 12.2x26" (31 x 66cm), clear, biohazard, printed, marking area, 200/cs
These high-temperature autoclavable bags are manufactured from heavy-gauge polypropylene material that withstands autoclaving at temperatures up to 134˚C (270˚F). Their ability to contain is completely unaffected at standard autoclave temperature (121˚C). Each bag has an oversized white marking panel, suitable for use with all types of permanent markers. They are also prominently marked with the words “Hazardous Waste” and the standard “Biohazard” symbol, conforming to the OSHA standard (29CFR1910.1030). Designed to resist stretching and poking, these bags are extremely pliable, leak-resistant and have seamless bottoms for exceptional strength and integrity. Both sizes (12×26 inch and 24×32 inch) are available in clear or opaque “biohazard” red color. They are supplied in units of 200 bags, with 4 inner packs of 50 bags for convenient dispensing and storage.
Features and Benefits
  • Heavy gauge polypropylene
  • Large white marking areas
  • Strong seamless bottom
  • Printed biohazard symbol
  • Withstands autoclaving at temperatures up to 134˚C (270˚F)
Legal Information
MTC is a trademark of MTC Bio
Still not finding the right product?
Explore all of our products under MTC Bio Autoclave Bags
Choose from one of the most recent versions:
Certificates of Analysis (COA)