Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSST0020

Sigma-Aldrich

SILuLite MAPK3 Mitogen activated protein kinase 3 human

recombinant, expressed in HEK 293 cells, MS Protein Standard

Synonym(s):

Erk1, MAP kinase 3, p44

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.12
Pricing and availability is not currently available.

recombinant

expressed in HEK 293 cells

Quality Level

Assay

≥95% (SDS-PAGE)

form

liquid

mol wt

45.5 kDa

suitability

suitable for mass spectrometry

UniProt accession no.

storage temp.

−20°C

Related Categories

General description

SILu Lite MAPK3 is a recombinant human protein expressed in human 293 cells. It is a monomer of 399 amino acids (including N-terminal polyhistidine and flag tags), with a molecular weight of ~45.5 kDa. SILu Lite MAPK3 is an analytical standard designed to be used as starting material for preparation of calibrators and controls in LC-MS applications.

Biochem/physiol Actions

MAPK3, the first mammalian MAPK to be characterized and cloned, is commonly expressed in most tissues and is activated through the small guanosine triphosphatase Ras and sequential activation of the protein kinases Raf and MEK upon stimulation of cells with a broad range of extracellular signals. Erk was suggested to be a good marker for estradiol and tamoxifen effects. Alzheimer′s disease′s Erk1 and Erk2 index values were inversely correlated with disease duration, suggesting maximal efficacy for early diagnosis. In addition, differential Erk1 and Erk2 phosphorylation, could have important clinical utility for providing increased certainty in the positive diagnosis of AD, particularly in the early phase of disease progression.

Sequence

MDYKDDDDKGHHHHHHHHGGQAAAAAQGGGGGEPRRTEGVGPGVPGEVEMVKGQPFDVGPRYTQLQYIGEGAYGMVSSAYDHVRKTRVAIKKISPFEHQTYCQRTLREIQILLRFRHENVIGIRDILRASTLEAMRDVYIVQDLMETDLYKLLKSQQLSNDHICYFLYQILRGLKYIHSANVLHRDLKPSNLLINTTCDLKICDFGLARIADPEHDHTGFLTEYVATRWYRAPEIMLNSKGYTKSIDIWSVGCILAEMLSNRPIFPGKHYLDQLNHILGILGSPSQEDLNCIINMKARNYLQSLPSKTKVAWAKLFPKSDSKALDLLDRMLTFNPNKRITVEEALAHPYLEQYYDPTDEPVAEEPFTFAMELDDLPKERLKELIFQETARFQPGVLEAP

Physical form

Supplied as a 0.1 mg/mL solution of 20mM sodium phosphate, pH 8.0, 1M NaCl.

Legal Information

This product is licensed under U.S. Patent No. 7,396,688 and foreign counterparts from E. I. du Pont de Nemours and Company. The purchase of this product conveys to the buyer the nontransferable right to use the purchased amount of the product for research and development only, including services for a third party for consideration. The buyer cannot sell or otherwise transfer this product, its components or materials made using this product or its components to a third party. Information about licenses for excluded uses is available from: E. I. du Pont de Nemours and Company; Attn: Associate Director, Commercial Development; DuPont Experimental Station E268; 200 Powdermill Rd.; Wilmington, DE 19803; 1-877-881-9787 (voice), 1-302-695-1437 (fax), [email protected].
SILu is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service