Skip to Content
MilliporeSigma

MSST0011

SILu™Prot ALB Albumin human

recombinant, expressed in HEK 293 cells, SIL MS Protein Standard, 13C and 15N-labeled

Synonym(s):

Albumin, LC/MS internal standard for human albumin in, stable isotope-labeled human albumin

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.12
UNSPSC Code:
23201100
Biological source:
human
Assay:
≥98% (SDS-PAGE)
Form:
lyophilized powder
Technique(s):
mass spectrometry (MS): suitable, protein mass spectrometry: suitable
Concentration:
≥10 μg/mg (Bradford)
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

human

Quality Segment

recombinant

expressed in HEK 293 cells

tag

His tagged

assay

≥98% (SDS-PAGE)

form

lyophilized powder

potency

≥98% Heavy amino acids incorporation efficiency by MS

concentration

≥10 μg/mg (Bradford)

technique(s)

mass spectrometry (MS): suitable, protein mass spectrometry: suitable

suitability

suitable for mass spectrometry (standard)

UniProt accession no.

storage temp.

−20°C

Gene Information

human ... ALB(213)

General description

SILu™Prot ALB is a recombinant, stable isotope-labeled human albumin which incorporates [13C6, 15N4]-Arginine and [13C6, 15N2]-Lysine. Expressed in human 293 cells, it is designed to be used as an internal standard for bioanalysis of albumin in mass-spectrometry. SILu™Prot ALB is a monomer of 607 amino acids (including C-terminal polyhistidine and flag tags), with a molecular weight of ~69 kDa.

Biochem/physiol Actions

Albumin is synthesized exclusively by the liver parenchymal cells at a rate of 14 g/day. While it is the most abundant plasma protein (>50%), increased urine albumin concentrations indicate bleedings in the lower urinary tract (ureter, bladder) or infections of the renal pelvis. A small abnormal albumin excretion is known as Microalbuminuria and serves as an indicator for temporary overload of the glomerular filtration (fever, excessive sports) or chronic injury of the glomeruli (diabetes).

Physical form

Supplied as a lyophilized powder containing phosphate buffered saline.

Other Notes

RRDAHKSEVAHRFKDLGEENFKALVLIAFAQYLQQCPFEDHVKLVNEVTEFAKTCVADESAENCDKSLHTLFGDKLCTVATLRETYGEMADCCAKQEPERNECFLQHKDDNPNLPRLVRPEVDVMCTAFHDNEETFLKKYLYEIARRHPYFYAPELLFFAKRYKAAFTECCQAADKAACLLPKLDELRDEGKASSAKQRLKCASLQKFGERAFKAWAVARLSQRFPKAEFAEVSKLVTDLTKVHTECCHGDLLECADDRADLAKYICENQDSISSKLKECCEKPLLEKSHCIAEVENDEMPADLPSLAADFVESKDVCKNYAEAKDVFLGMFLYEYARRHPDYSVVLLLRLAKTYETTLEKCCAAADPHECYAKVFDEFKPLVEEPQNLIKQNCELFEQLGEYKFQNALLVRYTKKVPQVSTPTLVEVSRNLGKVGSKCCKHPEAKRMPCAEDYLSVVLNQLCVLHEKTPVSDRVTKCCTESLVNRRPCFSALEVDETYVPKEFNAETFTFHADICTLSEKERQIKKQTALVELVKHKPKATKEQLKAVMDDFAAFVEKCCKADDKETCFAEEGKKLVAASQAALGLDYKDDDDKGHHHHHHHHGGQ

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC


Storage Class

11 - Combustible Solids

WGK

WGK 2

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library