Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MSPS001

Sigma-Aldrich

SILuPep Glucagon

synthetic, SIL MS Peptide Standard, ¹³C labeled

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100
NACRES:
NA.24

Quality Level

Assay

≥95% (HPLC)

form

lyophilized

color

off-white

shipped in

ambient

storage temp.

−20°C

Analysis Note

General Description
SILuPep Glucagon is a synthetic, stable isotope-labeled human peptide.

Sequence
HSQGTFTSDYSKY-Leu(U13C6)-DSRRAQDFVQW-Leu(U13C6)-MNT
Terminal modifications: N-Term: [H] C-Term: [OH]
Length (residues): 29

Label Incorporation
>= 98% as determined by mass spectrometry

Legal Information

SILu is a trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1A

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service