Skip to Content
MilliporeSigma

ML0012

Pentose Phosphate Metabolite Library

Synonym(s):

PPP Metabolite Library

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.24
UNSPSC Code:
41116107
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


application(s)

advanced drug delivery

storage temp.

−20°C

Other Notes

Contains up to 10 mg each of the pentose phosphate pathway metabolite standards packaged individually.
For additional information on our range of Biochemicals, please complete this form.


Kit Components Also Available Separately

Product No.
Description
SDS & Pricing

  • D-Xylulose 5-phosphate lithium salt, ≥90% (TLC) 10 mg
    SDS

  • D-Sedoheptulose 7-phosphate lithium salt, ≥90% (TLC) 1 mg
    SDS

  • 2-Deoxyribose 5-phosphate sodium salt, ≥95% (ICP-AES) 10 mg
    SDS

  • Dihydroxyacetone phosphate dilithium salt, ≥93% dry basis (enzymatic) 10 mg
    SDS

  • D-Erythrose 4-phosphate sodium salt, ≥50% (TLC) 10 mg
    SDS

  • D-Fructose 6-phosphate disodium salt hydrate, ≥98%, amorphous powder 10 mg
    SDS

  • D-Fructose 1,6-bisphosphate trisodium salt hydrate, ≥98% (TLC) 10 mg
    SDS

See All (13)

pictograms

Exclamation mark

signalword

Warning

Storage Class

11 - Combustible Solids

wgk

WGK 3

hcodes

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library