Skip to Content
MilliporeSigma

MAS004

L-Glutamate Quick Test Strips

Sufficient for 10 tests

Synonym(s):

ʟ-Glutamic Acid Quick Test Strips

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12352106
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


input

beverage(s)
food(s)

application(s)

food and beverages

detection method

colorimetric

storage temp.

2-8°C

General description

L-Glutamic acid (L-glutamate), one of the 20 common amino acids, is naturally present in various foods, including cheese, milk, meat, fish, corn, tomatoes, mushrooms, soybeans, and sugar beets. It is also found in processed fruit and vegetables like tomato puree, ketchup, and soy sauce, as well as in condiments, processed meat products such as extracts, bouillon, and sausages, soups, pharmaceuticals, and other materials, including biological cultures and samples. In its free form, L-glutamic acid serves as a major flavor enhancer in foods with a meaty or savory taste. Its role in enhancing the palatability of dishes is utilized through traditional cooking methods that release free L-glutamic acid or through supplementation with monosodium glutamate (MSG).Excessive use of MSG, an E621 food additive commonly found in processed foods, can lead to symptoms like palpitations, severe headaches, nausea, wheezing, and increased body temperature, which are believed to result from the neuro-excitatory properties of free L-glutamic acid. Additionally, glutamate plays a crucial role as a mammalian neurotransmitter and is associated with various neurological and psychiatric disorders, including lateral sclerosis, autism, and Alzheimer′s disease. Furthermore, it functions as a metabolite in cell cultures, where maintaining the glutamate concentration within an appropriate range is essential for optimal cell growth and product formation. For the precise and rapid determination of L-Glutamate, which includes monosodium glutamate or MSG, in food and beverage samples as well as biological samples, Sigma-Aldrich′s L-Glutamate Quick Test Strips provide a simple, reliable, and direct method.

Application

The L-Glutamate Quick Test Strips can be used for determining L-Glutamate levels in:
  • Food and Beverage Testing
  • Metabolomics Research
This kit is suitable for the detection of L-glutamate in a variety of food and beverage samples.

Biochem/physiol Actions

Glutamate Quick Test Strips are based on glutamate dehydrogenase catalyzed oxidation of glutamate in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glutamate concentration in the sample.
The L-Glutamate Quick Test Strips are based on glutamate dehydrogenase catalyzed oxidation of glutamate in which the formed NADH reduces a chromogenic reagent. The intensity of the product color is directly proportional to the glutamate concentration in the sample.

Features and Benefits

  • Fast and Sensitive: Offers rapid results with the flexibility to use 20 or 100 µL of sample for semi-quantitative measurement between 0-300 mg/L (undiluted) L-Glutamate
  • Quick Results: Sample treatment and assay can be completed in under 15 minutes, ensuring efficiency
  • User-Friendly Design: Convenient test strip design eliminates the requirement for a plate or cuvette reader

Other Notes

For additional information on our range of Biochemicals, please complete this form.


Storage Class

10 - Combustible liquids



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library