Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK453

Sigma-Aldrich

Glutathione S-transferase Assay Kit

Sufficient for 100 colorimetric tests

Synonym(s):

GST Assay Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

input

cell lysate
tissue

detection method

colorimetric

relevant disease(s)

neurological disorders, asthma; pulmonary disorders, diabetes; cancer; endocrinological disorders

storage temp.

−20°C

General description

Glutathione S-transferase (GST) is a vital enzyme involved in detoxification, aiding in the removal of harmful substances from cells. It plays a key role in metabolizing a wide range of compounds, including drugs, toxins, and carcinogens, and contributes to cellular processes such as signaling and inflammation regulation. Monitoring GST levels is crucial for several reasons: it serves as a biomarker for cellular stress, indicating damage from toxins or oxidative stress; it aids in diagnosing and managing various diseases, including cancer and liver diseases; it informs pharmacological studies by assessing drug efficacy and toxicity; and it facilitates environmental monitoring, identifying pollution and potential health risks. Overall, monitoring GST levels provides valuable insights into cellular health, disease status, drug metabolism, and environmental exposure, guiding early detection and management of health conditions.

Application

The Glutathione S-transferase Assay Kit may be used for:
  • Cancer Research
  • Oxidative Stress Research
  • Metabolism Research

Features and Benefits

  • Efficient and Precise: Requires only 20 μL of sample and a quick 10-minute reaction time while detecting Glutathione S-transferase levels within a range of 2 to 80 U/L, offering convenience and sensitivity.
  • Compatibility with High-Throughput Systems: Easily integrate our kit into high-throughput handling systems, ensuring smooth operation and accurate results, enhancing efficiency in your laboratory processes.

Suitability

The kit is suitable for Glutathione S-transferase activity determination in cell lysates, tissues, etc.

Principle

The Glutathione S-transferase Assay Kit is based on the GST enzyme reaction between GSH and the GST substrate, CDNB (1- chloro-2,4-dinitrobenzene). The GST catalyzed formation of GS-DNB produces a dinitrophenyl thioether which can be detected spectrophotometrically at A340 nm. The rate of increase in absorbance at A340 nm is directly proportional to the GST activity in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Irrit. 2 - Skin Sens. 1

Storage Class Code

10 - Combustible liquids

Flash Point(F)

188.6 °F

Flash Point(C)

87 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service