Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK445

Sigma-Aldrich

Protein Creatinine Ratio Assay Kit

Sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

storage condition

(No metal containers. Tightly closed.)

input

urine

detection method

colorimetric

relevant disease(s)

endocrinological disorders, diabetes; cardiovascular diseases, hypertension

storage temp.

2-8°C

General description

Protein is filtered out of urine by the glomeruli of the kidneys. Albumin is the most common serum protein, thus the majority of the protein in urine is albumin. A damaged kidney will allow some protein through into the urine, the less protein in urine the better. Elevated protein levels in urine is called microalbuminuria or proteinuria, which typically arises due to type 1 diabetes, type 2 diabetes, or high blood pressure.

Creatinine is synthesized in the body at a fairly constant rate from creatine. In healthy individuals, creatinine secretion is independent of diet and is fairly constant. The creatinine clearance test has become one of the most sensitive tests for measuring glomerular filtration rate. The Protein/Creatinine Ratio stands out as the simplest and most convenient test for detecting proteinuria/microalbuminuria.

Application

The Protein Creatinine Ratio Assay Kit may be used for:
  • Measuring Glomerular Filtration Rate
  • Renal Metabolism Research

Features and Benefits

  • Enhanced Sensitivity and Accuracy: Detect protein levels ranging from 1 to 20 mg/dL and creatinine levels ranging from 1 to 150 mg/dL in a linear fashion within a 96-well plate, providing precise results.
  • Efficient and Rapid: Experience a streamlined process with no sample pre-treatment required, enabling a straightforward add-incubate-read procedure that takes just 10 minutes to complete, saving valuable time.
  • Compatibility with High-Throughput Systems: Easily integrate our kit into high-throughput handling systems, ensuring seamless operations and maintaining efficiency and accuracy in your laboratory workflow.

Suitability

The kit is suitable for the quantitative determination of urinary protein and creatinine concentrations (protein/creatinine ratio, UPCR) in urine samples.

Principle

Protein/Creatinine Ratio (PCR) remains the simplest and most convenient test for proteinuria. Other methods such as the 24-hour urine test or timed urine test require strict adherence to sample collection protocol. Since the protein concentration is normalized to creatinine secretion, the urine sample can be taken at any time and no diet or liquid restrictions are necessary for sample collection.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Met. Corr. 1 - Skin Irrit. 2 - STOT SE 2

Target Organs

Eyes,Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service