Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

MAK434

Sigma-Aldrich

Acetaldehyde Assay Kit

Sufficient for 100 colorimetric tests

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

Quality Level

input

cell culture medium
tissue
serum
urine
food(s)
beverage(s)
plasma

application(s)

agriculture
cosmetics
environmental
food and beverages

detection method

colorimetric

relevant disease(s)

gastrointestinal diseases

storage temp.

−20°C

General description

Acetaldehyde (CH₃CHO) is a commonly found aldehyde in nature and widely used in various industries. It is a metabolic byproduct of ethanol in the liver and is toxic to the human body. However, it is rapidly converted to the less harmful acetic acid by the enzyme aldehyde dehydrogenase. Individuals deficient in aldehyde dehydrogenase experience accumulation of acetaldehyde upon alcohol consumption, leading to facial and body flushing known as alcohol flush reaction or “Asian flush syndrome.” Acetaldehyde buildup has also been linked to alcohol-related hangover effects. Despite its classification as a carcinogen, acetaldehyde is naturally present in many foods and beverages such as ripe fruit, coffee, and wine.

Application

The Acetaldehyde Assay Kit may be used for:
  • Alcohol Metabolism Research
  • Hepatology Research
  • Neurological Research

Features and Benefits

  • Simplified Process: Enjoy a hassle-free procedure that requires the addition of a single working reagent and only a 30 minute assay room temperature reaction, saving you time and effort.
  • Convenient and Sensitive: Uses only 20 µL of sample for the determination of acetaldehyde with a detection range of 2 µM to 2mM.
  • High-Throughput Compatibility: Seamlessly integrate our kit into high-throughput handling systems, ensuring efficiency and accuracy.

Suitability

The kit is suitable for acetaldehyde determination in biological samples (e.g. plasma, serum, urine, tissue and culture media) and food/beverage samples (e.g. wine, coffee, and juice).

Principle

The Acetaldehyde Assay Kit is based on the aldehyde dehydrogenase catalyzed oxidation of acetaldehyde with the reduction of NAD to NADH. The formed NADH subsequently reduces MTT, producing a colored formazan compound. The intensity of the end product color, measured at 565 nm, is directly proportional to the acetaldehyde concentration in the sample.

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

12 - Non Combustible Liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service