Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK167

Sigma-Aldrich

Fluorometric Maleimide Assay Kit

sufficient for 200 fluorometric tests

Synonym(s):

Maleimide Fluorescence Detection Kit

Sign Into View Organizational & Contract Pricing


About This Item

EC Number:
UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 200 fluorometric tests

detection method

fluorometric

relevant disease(s)

endocrinological disorders, diabetes

storage temp.

−20°C

General description

Maleimide is an unsaturated imide used in protein conjugation reactions.

Suitability

The assay can be performed in either a 96 or 384 multiwell plate format using a fluorescence microplate reader.

Principle

The Fluorometric Maleimide Assay Kit provides a sensitive, one-step fluorometric assay to detect maleimide, which reacts with a proprietary dye, resulting in a fluorometric product (λex = 490/λem = 520 nm) proportional to the amount of maleimide present. The assay can be performed in either a 96 or 384 multiwell plate format using a fluorescence microplate reader.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service