Skip to Content
MilliporeSigma
All Photos(2)

Key Documents

MAK161

Sigma-Aldrich

Multidrug Resistance Assay Kit

(Fluorometric MDR Assay)

Synonym(s):

MDR Assay

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

(Fluorometric MDR Assay)

application(s)

pharmaceutical

detection method

fluorometric

relevant disease(s)

cancer

storage temp.

−20°C

Gene Information

General description

Acquired resistance to chemotherapy drugs, multidrug resistance or MDR, is a major contributor to treatment failure for many types of cancers. MDR is typically associated with the increased expression of two ATP-dependent drug efflux pumps, P-Glycoprotein (P-gp or MDR1) and the Multidrug Resistance-Associated Protein (MRP1). These pumps actively expel chemotherapeutic agents, typically hydrophobic amphipathic natural products, from the cytoplasm to exterior of the cell.

This kit is utilizes a hydrophobic fluorescent dye molecule to assess MDR activity in cells. This dye rapidly penetrates cell membranes and becomes trapped resulting in an increase in fluorescence intensity (λex = 490/λem = 525 nm). In cells expressing MDR transporters, the dye is rapidly extruded by the transporters, resulting in decreased fluorescence intensity.

Features and Benefits

Compatible with high-throughput handling systems.

Suitability

This kit is suitable for the screening of MDR pump inhibitors or for identifying cell lines with high MDR activity.

Principle

This kit is utilizes a hydrophobic fluorescent dye molecule to assess MDR activity in cells. This dye rapidly penetrates cell membranes and becomes trapped resulting in an increase in fluorescence intensity (λEx = 490/λEm = 525 nm). In cells expressing MDR transporters, the dye is rapidly extruded by the transporters, resulting in decreased fluorescence intensity.

Storage Class Code

10 - Combustible liquids


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Drug resistance in cancer: an overview.
Housman G, et al.
Cancer, 6(3), 1769-1792 (2014)
Mechanisms and insights into drug resistance in cancer.
Zahreddine H and Katherine B
Frontiers in Pharmacology, 4, 28-28 (2013)
Non-coding polymorphisms in nucleotide binding domain 1 in ABCC1 gene associate with transcript level and survival of patients with breast cancer.
Kunicka T, et al.
PLoS ONE, 9(7), e101740-e101740 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service