Sign In to View Organizational & Contract Pricing.
Select a Size
Change View
About This Item
UNSPSC Code:
12161503
EC Number:
200-664-3
NACRES:
NA.84
usage
sufficient for 100 fluorometric tests
detection method
fluorometric
relevant disease(s)
endocrinological disorders, diabetes; gastrointestinal diseases; cancer
storage temp.
−20°C
General description
Fatty acids are critical for many biological processes including energy metabolism and the synthesis of structural cellular components. The facilitated uptake of fatty acids, presumably through fatty acid transport proteins, is thought to be regulated at multiple levels. The dysregulation of fatty acid uptake may contribute to diseases such as obesity, type 2 diabetes, and hepatic steatosis.
Application
This kit is suitable for the measurement of fatty acid uptake in cells containing fatty acid transporters
Biochem/physiol Actions
This kit is provides a simple and sensitive method for the measurement of fatty acid uptake in cells containing fatty acid transporters. This kit uses a proprietary dodecanoic acid fluorescent fatty acid substrate whose uptake into cells result in an increase in fluorescence intensity (λex = 485/λem = 515 nm).
Storage Class
10 - Combustible liquids
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.