Skip to Content
MilliporeSigma

MAK129

β-Glucosidase Activity Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Beta-Glucosidase Assay Kit

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

NACRES:
NA.84
UNSPSC Code:
12161503
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

neurological disorders; gastrointestinal diseases; hematological disorder

storage temp.

−20°C

General description

β-Glucosidase hydrolyzes carbohydrates by acting on terminal, non-reducing b(1→4)-linked D-glucose residues with the release of D-glucose. β-Glucosidases are required by organisms for the consumption of cellulose. Lysozyme, a β-glucosidase present in tears, acts on the β(1→4) glucose bonds present in the peptidoglycan cell wall of Gram-negative bacteria and helps to prevent bacterial infections in the eye. Defects in β-glucosidase activity have been implicated in Gaucher′s disease and Parkinson′s disease.

Application

β-Glucosidase Activity Assay Kit has been used to measure the β-glucosidase activity of the sample.
The β-Glucosidase Activity Assay kit provides a simple and direct procedure for measuring β-glucosidase activity in biological samples, for characterizing β-glucosidase production, and for the screening of β-glucosidase modulators.

Biochem/physiol Actions

In this assay, β-glucosidase activity is determined by a reaction in which β-glucosidase hydrolyzes p-nitrophenyl-β-D-glucopyranoside resulting in the formation of a colorimetric (405 nm) product, proportional to the β-glucosidase activity present. One unit of β-Glucosidse is the amount of enzyme that catalyzes the hydrolysis of 1.0 μmole substrate per minute at pH 7.0.

Features and Benefits

Compatible with high-throughput handling systems.


Storage Class

12 - Non Combustible Liquids

wgk

WGK 2



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library